2,3-Difluoro-4-methoxybenzamide

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CA-4401-5G CA-4401-1G
COBBCA-4401-5GEA 0 GBP
COBBCA-4401-5G COBBCA-4401-1G
2,3-Difluoro-4-methoxybenzamide
2,3-Difluoro-4-methoxybenzamide
Formula: C₈H₇F₂NO₂
MDL Number: MFCD04115983
CAS Number: 886500-67-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR