3-Ethoxy-2-fluorobenzonitrile

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OT-0032-5G OT-0032-1G
COBBOT-0032-5GEA 0 GBP
COBBOT-0032-5G COBBOT-0032-1G
3-Ethoxy-2-fluorobenzonitrile
3-Ethoxy-2-fluorobenzonitrile
Formula: C₉H₈FNO
MDL Number: MFCD10699689
CAS Number: 1033202-20-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR