(2-Methyl-3-nitrophenyl)acetic acid
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2-Methyl-3-nitrophenylacetic Acid
AS21769G100
AS21769G25
ACTIAS21769G100EA
0
GBP
ACTIAS21769G100
ACTIAS21769G25
(2-Methyl-3-nitrophenyl)acetic acid
(2-Methyl-3-nitrophenyl)acetic acid
Formula:
C₉H₉NO₄ MW: 195.17 g/mol |
MDL Number:
MFCD07782132 CAS Number: 23876-15-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...