2,4-Dichloro-5-fluorobenzamide

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

005728-5G 005728-1G
FLUO005728-5GEA 0 GBP
FLUO005728-5G FLUO005728-1G
2,4-Dichloro-5-fluorobenzamide
2,4-Dichloro-5-fluorobenzamide
Formula: C₇H₄Cl₂FNO
MW: 208.02 g/mol
MDL Number: MFCD01631478
CAS Number: 135748-33-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR