(±)-α-Chlorophenylacetic acid
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CA-4975-5G
CA-4975-1G
COBBCA-4975-5GEA
0
GBP
COBBCA-4975-5G
COBBCA-4975-1G
(±)-α-Chlorophenylacetic acid
(±)-α-Chlorophenylacetic acid
Formula:
C₈H₇ClO₂ |
MDL Number:
MFCD00037753 CAS Number: 4755-72-0 EINECS: 225-284-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...