5-(2,4-Dichlorophenyl)-3-(3-nitrophenyl)-1,2,4-oxadiazole

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-6601-1G HC-6601-5G
COBBHC-6601-1GEA 0 GBP
COBBHC-6601-1G COBBHC-6601-5G
5-(2,4-Dichlorophenyl)-3-(3-nitrophenyl)-1,2,4-oxadiazole
5-(2,4-Dichlorophenyl)-3-(3-nitrophenyl)-1,2,4-oxadiazole
Formula: C₁₄H₇Cl₂N₃O₃
MDL Number: MFCD03717917
CAS Number: 694521-58-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR