3-(4-Bromophenyl)-5-(2,4-dichlorophenyl)-1,2,4-oxadiazole

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-6599-5G HC-6599-1G
COBBHC-6599-5GEA 0 GBP
COBBHC-6599-5G COBBHC-6599-1G
3-(4-Bromophenyl)-5-(2,4-dichlorophenyl)-1,2,4-oxadiazole
3-(4-Bromophenyl)-5-(2,4-dichlorophenyl)-1,2,4-oxadiazole
Formula: C₁₄H₇BrCl₂N₂O
MDL Number: MFCD02217937
CAS Number: 381178-19-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR