5-Methyl-3-(4-methylphenyl)-1,2,4-oxadiazole

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 5-Methyl-3-p-tolyl-1,2,4-oxadiazole

HC-6424-1G HC-6424-5G
COBBHC-6424-1GEA 0 GBP
COBBHC-6424-1G COBBHC-6424-5G
5-Methyl-3-(4-methylphenyl)-1,2,4-oxadiazole
5-Methyl-3-(4-methylphenyl)-1,2,4-oxadiazole
Formula: C₁₀H₁₀N₂O
MDL Number: MFCD00085801
CAS Number: 81386-30-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR