2-(Benzylamino)nicotinonitrile

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PY-7982-1G PY-7982-5G
COBBPY-7982-1GEA 0 GBP
COBBPY-7982-1G COBBPY-7982-5G
2-(Benzylamino)nicotinonitrile
2-(Benzylamino)nicotinonitrile
Formula: C₁₃H₁₁N₃
MDL Number: MFCD09455270
CAS Number: 50351-72-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR