2-Phenylisonicotinonitrile

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

3Y-0714.1 3Y-0714.5 3Y-0714.500
BIOT3Y-0714.1EA 0 GBP
BIOT3Y-0714.1 BIOT3Y-0714.5 BIOT3Y-0714.500
2-Phenylisonicotinonitrile
2-Phenylisonicotinonitrile 2-(4-Formylphenyl)isonicotinonitrile
Formula: C₁₂H₈N₂
MDL Number: MFCD06200958
CAS Number: 33744-17-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR