2-(Benzylamino)-5-bromo-3-nitropyridine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PY-7364-5G PY-7364-1G
COBBPY-7364-5GEA 0 GBP
COBBPY-7364-5G COBBPY-7364-1G
2-(Benzylamino)-5-bromo-3-nitropyridine
2-(Benzylamino)-5-bromo-3-nitropyridine
Formula: C₁₂H₁₀BrN₃O₂
MDL Number: MFCD09862702
CAS Number: 954228-85-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR