2-Acetamidoisonicotinic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CA-4040-5G CA-4040-25G CA-4040-1G
COBBCA-4040-5GEA 0 GBP
COBBCA-4040-5G COBBCA-4040-25G COBBCA-4040-1G
2-Acetamidoisonicotinic acid
2-Acetamidoisonicotinic acid
Formula: C₈H₈N₂O₃
MDL Number: MFCD00233713
CAS Number: 54221-95-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR