Methyl-2-chloro-6-methylisonicotinate

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS22207G25 AS22207G5
ACTIAS22207G25EA 0 GBP
ACTIAS22207G25 ACTIAS22207G5
Methyl-2-chloro-6-methylisonicotinate
Methyl 2-chloro-6-methylisonicotinate
Formula: C₈H₈ClNO₂
MDL Number: MFCD04038391
CAS Number: 3998-90-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR