2-Amino-5-bromo-4-methyl-3-nitropyridine
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OT-0051-5G
OT-0051-1G
OT-0051-25G
COBBOT-0051-5GEA
0
GBP
COBBOT-0051-5G
COBBOT-0051-1G
COBBOT-0051-25G
2-Amino-5-bromo-4-methyl-3-nitropyridine
2-Amino-5-bromo-4-methyl-3-nitropyridine
Formula:
C₆H₆BrN₃O₂ MW: 232.0400 g/mol |
MDL Number:
MFCD00092197 CAS Number: 100367-40-6 |
Learn more
![](/m-uk.vwr.com/en_GB/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...