2-Acetamido-5-bromoisonicotinic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CA-4095-5G CA-4095-1G
COBBCA-4095-5GEA 0 GBP
COBBCA-4095-5G COBBCA-4095-1G
2-Acetamido-5-bromoisonicotinic acid
2-Acetamido-5-bromoisonicotinic acid
Formula: C₈H₇BrN₂O₃
MDL Number: MFCD07363802
CAS Number: 871269-03-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR