2-Methoxyisonicotinic acid hydrazide

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PY-7031-1G PY-7031-5G
COBBPY-7031-1GEA 0 GBP
COBBPY-7031-1G COBBPY-7031-5G
2-Methoxyisonicotinic acid hydrazide
2-Methoxyisonicotinic acid hydrazide
Formula: C₇H₉N₃O₂
MDL Number: MFCD01747192
CAS Number: 19353-97-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR