5-Bromo-2-(cyclopentylamino)-3-nitropyridine
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PY-7365-5G
PY-7365-1G
COBBPY-7365-5GEA
0
GBP
COBBPY-7365-5G
COBBPY-7365-1G
5-Bromo-2-(cyclopentylamino)-3-nitropyridine
5-Bromo-2-(cyclopentylamino)-3-nitropyridine
Formula:
C₁₀H₁₂BrN₃O₂ |
MDL Number:
MFCD08692161 CAS Number: 1033202-53-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...