5-Bromo-2-(cyclohexylamino)-3-nitropyridine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PY-7334-1G PY-7334-5G
COBBPY-7334-1GEA 0 GBP
COBBPY-7334-1G COBBPY-7334-5G
5-Bromo-2-(cyclohexylamino)-3-nitropyridine
5-Bromo-2-(cyclohexylamino)-3-nitropyridine
Formula: C₁₁H₁₄BrN₃O₂
MDL Number: MFCD08692170
CAS Number: 1033202-38-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR