5-Bromo-2-(cyclohexylamino)pyridine
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PY-7994-1G
PY-7994-5G
COBBPY-7994-1GEA
0
GBP
COBBPY-7994-1G
COBBPY-7994-5G
5-Bromo-2-(cyclohexylamino)pyridine
5-Bromo-2-(cyclohexylamino)pyridine
Formula:
C₁₁H₁₅BrN₂ |
MDL Number:
MFCD11124008 CAS Number: 942050-72-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...