2-Bromo-6-(cyclohexylamino)pyridine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-2723-5G HC-2723-1G
COBBHC-2723-5GEA 0 GBP
COBBHC-2723-5G COBBHC-2723-1G
2-Bromo-6-(cyclohexylamino)pyridine
2-Bromo-6-(cyclohexylamino)pyridine
Formula: C₁₁H₁₅BrN₂
MDL Number: MFCD09878953
CAS Number: 959237-36-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR