5-Bromo-2-(isopropylamino)-3-nitropyridine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PY-7336-5G PY-7336-1G
COBBPY-7336-5GEA 0 GBP
COBBPY-7336-5G COBBPY-7336-1G
5-Bromo-2-(isopropylamino)-3-nitropyridine
5-Bromo-2-(isopropylamino)-3-nitropyridine
Formula: C₈H₁₀BrN₃O₂
MDL Number: MFCD08692141
CAS Number: 954228-62-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR