2-(N-Benzyl-N-methylamino)-5-bromopyridine
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PY-1134-5G
PY-1134-1G
COBBPY-1134-5GEA
0
GBP
COBBPY-1134-5G
COBBPY-1134-1G
2-(N-Benzyl-N-methylamino)-5-bromopyridine
2-(N-Benzyl-N-methylamino)-5-bromopyridine
Formula:
C₁₃H₁₃BrN₂ MW: 263.14 g/mol |
MDL Number:
MFCD12913988 CAS Number: 1187386-36-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...