6-Bromo-2-(diethylamino)pyridine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-6365-25G HC-6365-1G HC-6365-5G
COBBHC-6365-25GEA 0 GBP
COBBHC-6365-25G COBBHC-6365-1G COBBHC-6365-5G
6-Bromo-2-(diethylamino)pyridine
6-Bromo-2-(diethylamino)pyridine
Formula: C₉H₁₃BrN₂
MDL Number: MFCD09878973
CAS Number: 856850-57-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR