2-Bromoisonicotinic acid hydrazide

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PY-7245-1G PY-7245-25G PY-7245-5G
COBBPY-7245-1GEA 0 GBP
COBBPY-7245-1G COBBPY-7245-25G COBBPY-7245-5G
2-Bromoisonicotinic acid hydrazide
2-Bromoisonicotinic acid hydrazide
Formula: C₆H₆BrN₃O
MDL Number: MFCD04039856
CAS Number: 29849-15-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR