2-Bromo-N-methylisonicotinamide

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PY-7227-1G PY-7227-25G PY-7227-5G
COBBPY-7227-1GEA 0 GBP
COBBPY-7227-1G COBBPY-7227-25G COBBPY-7227-5G
2-Bromo-N-methylisonicotinamide
2-Bromo-N-methylisonicotinamide
Formula: C₇H₇BrN₂O
MDL Number: MFCD03411643
CAS Number: 337536-01-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR