2-(N-Boc-N-methylamino)-5-bromopyridine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: tert-Butyl (5-bromopyridin-2-yl)(methyl)carbamate

PY-1120-5G PY-1120-1G
COBBPY-1120-5GEA 0 GBP
COBBPY-1120-5G COBBPY-1120-1G
2-(N-Boc-N-methylamino)-5-bromopyridine
2-(N-Boc-N-methylamino)-5-bromopyridine
Formula: C₁₁H₁₅BrN₂O₂
MW: 287.1530 g/mol
MDL Number: MFCD12546613
CAS Number: 227939-01-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR