5-Bromo-2-chloro-3-hydroxypyridine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 5-Bromo-2-chloropyridin-3-ol

AS36472G1 AS36472G5
ACTIAS36472G1EA 44 GBP
ACTIAS36472G1 ACTIAS36472G5
5-Bromo-2-chloro-3-hydroxypyridine
5-Bromo-2-chloro-3-hydroxypyridine
Formula: C₅H₃BrClNO
MW: 208.4400 g/mol
MDL Number: MFCD08688592
CAS Number: 286946-77-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR