4-Pyridineboronic acid hydrochloride

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Pyridin-4-ylboronic acid hydrochloride

017173-1G 017173-25G 017173-5G
FLUO017173-1GEA 0 GBP
FLUO017173-1G FLUO017173-25G FLUO017173-5G
4-Pyridineboronic acid hydrochloride
4-Pyridineboronic acid hydrochloride
Formula: C₅H₇BClNO₂
MW: 159.38 g/mol
MDL Number: MFCD06201888
CAS Number: 913835-65-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR