4-Cyano-2-pyridineboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BB-5138-1G BB-5138-250M
COBBBB-5138-1GEA 0 GBP
COBBBB-5138-1G COBBBB-5138-250M
4-Cyano-2-pyridineboronic acid
4-Cyano-2-pyridineboronic acid
Formula: C₆H₅BN₂O₂
MDL Number: MFCD08703250
CAS Number: 1072946-01-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR