2-(Cyclohexyl(methyl)amino)-5-pyridineboronic acid pinacol ester
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PN-8767-1G
PN-8767-250M
COBBPN-8767-1GEA
0
GBP
COBBPN-8767-1G
COBBPN-8767-250M
2-(Cyclohexyl(methyl)amino)-5-pyridineboronic acid pinacol ester
2-(Cyclohexyl(methyl)amino)-5-pyridineboronic acid pinacol ester
Formula:
C₁₈H₂₉BN₂O₂ |
MDL Number:
MFCD09027071 CAS Number: 1073354-73-0 |
Learn more
![](/m-uk.vwr.com/en_GB/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...