3-Cyano-2-ethoxy-5-pyridineboronic acid pinacol ester
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PN-5869-1G
PN-5869-250M
COBBPN-5869-1GEA
0
GBP
COBBPN-5869-1G
COBBPN-5869-250M
3-Cyano-2-ethoxy-5-pyridineboronic acid pinacol ester
3-Cyano-2-ethoxy-5-pyridineboronic acid pinacol ester
Formula:
C₁₄H₁₉BN₂O₃ |
MDL Number:
MFCD12546500 CAS Number: 1218791-35-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...