6-Fluoro-3-isoquinolinol

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-6670-1G HC-6670-5G
COBBHC-6670-1GEA 0 GBP
COBBHC-6670-1G COBBHC-6670-5G
6-Fluoro-3-isoquinolinol
6-Fluoro-3-isoquinolinol
Formula: C₉H₆FNO
MDL Number: MFCD11053873
CAS Number: 51463-15-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR