5-Acetamidoisoquinoline

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AN-1515-5G AN-1515-1G
COBBAN-1515-5GEA 0 GBP
COBBAN-1515-5G COBBAN-1515-1G
5-Acetamidoisoquinoline
5-Acetamidoisoquinoline
Formula: C₁₁H₁₀N₂O
MDL Number: MFCD05981994
CAS Number: 27461-33-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR