6-Bromo-3-isoquinolinol

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 6-Bromo-3-hydroxyisoquinoline

HC-2057-5G HC-2057-1G
COBBHC-2057-5GEA 0 GBP
COBBHC-2057-5G COBBHC-2057-1G
6-Bromo-3-isoquinolinol
6-Bromo-3-isoquinolinol
Formula: C₉H₆BrNO
MW: 224.0500 g/mol
MDL Number: MFCD11110423
CAS Number: 1031927-91-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR