5-Bromo-8-nitroisoquinoline

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS20250G1 AS20250G25 AS20250G5
ACTIAS20250G1EA 0 GBP
ACTIAS20250G1 ACTIAS20250G25 ACTIAS20250G5
5-Bromo-8-nitroisoquinoline
5-Bromo-8-nitroisoquinoline
Formula: C₉H₅BrN₂O₂
MDL Number: MFCD02091227
CAS Number: 63927-23-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR