8-Amino-5-bromoisoquinoline

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 5-Bromoisoquinolin-8-amine

HC-2135-1G HC-2135-5G
COBBHC-2135-1GEA 0 GBP
COBBHC-2135-1G COBBHC-2135-5G
8-Amino-5-bromoisoquinoline
8-Amino-5-bromoisoquinoline
Formula: C₉H₇BrN₂
MW: 223.0693 g/mol
MDL Number: MFCD09261047
CAS Number: 90721-35-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR