Diphenyliodonium trifluoromethanesulphonate
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
009166-1G
009166-5G
FLUO009166-1GEA
0
GBP
FLUO009166-1G
FLUO009166-5G
Diphenyliodonium trifluoromethanesulphonate
Diphenyliodonium trifluoromethanesulphonate
Formula:
C₁₃H₁₀F₃IO₃S MW: 430.19 g/mol |
MDL Number:
MFCD00191356 CAS Number: 66003-76-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...