3-(Cyclohexylamino)propionitrile

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

018475-10G 018475-25G
FLUO018475-10GEA 0 GBP
FLUO018475-10G FLUO018475-25G
3-(Cyclohexylamino)propionitrile
3-(Cyclohexylamino)propionitrile
Formula: C₉H₁₆N₂
MDL Number: MFCD00021325
CAS Number: 702-03-4
EINECS: 211-864-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR