4-Hydroxy-6-methoxy-2-(trifluoromethyl)quinoline

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-2400-5G HC-2400-1G
COBBHC-2400-5GEA 0 GBP
COBBHC-2400-5G COBBHC-2400-1G
4-Hydroxy-6-methoxy-2-(trifluoromethyl)quinoline
4-Hydroxy-6-methoxy-2-(trifluoromethyl)quinoline
Formula: C₁₁H₈F₃NO₂
MDL Number: MFCD11855897
CAS Number: 1701-21-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR