3-Bromo-6-fluoro-4-hydroxy-2-(trifluoromethyl)quinoline

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-Bromo-6-fluoro-2-(trifluoromethyl)quinolin-4-ol

HC-2163-1G HC-2163-5G
COBBHC-2163-1GEA 0 GBP
COBBHC-2163-1G COBBHC-2163-5G
3-Bromo-6-fluoro-4-hydroxy-2-(trifluoromethyl)quinoline
3-Bromo-6-fluoro-4-hydroxy-2-(trifluoromethyl)quinoline
Formula: C₁₀H₄BrF₄NO
MW: 310.425 g/mol
MDL Number: MFCD11504911
CAS Number: 1072944-66-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR