7-Chloro-3-nitro-4-quinolinol

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 7-Chloro-3-nitroquinolin-4-ol

AS24531G1 AS24531G5
ACTIAS24531G1EA 0 GBP
ACTIAS24531G1 ACTIAS24531G5
7-Chloro-3-nitro-4-quinolinol
7-Chloro-3-nitro-4-quinolinol
Formula: C₉H₅ClN₂O₃
MW: 224.6006 g/mol
MDL Number: MFCD09800417
CAS Number: 5350-50-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR