6-Chloro-5-nitroquinoline

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-2109-5G HC-2109-1G
COBBHC-2109-5GEA 0 GBP
COBBHC-2109-5G COBBHC-2109-1G
6-Chloro-5-nitroquinoline
6-Chloro-5-nitroquinoline
Formula: C₉H₅ClN₂O₂
MW: 208.6012 g/mol
MDL Number: MFCD00093616
CAS Number: 86984-32-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR