4,6-Dichloroquinoline-3-carboxylic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PY-7874-1G PY-7874-5G
COBBPY-7874-1GEA 0 GBP
COBBPY-7874-1G COBBPY-7874-5G
4,6-Dichloroquinoline-3-carboxylic acid
4,6-Dichloroquinoline-3-carboxylic acid
Formula: C₁₀H₅Cl₂NO₂
MW: 242.582 g/mol
MDL Number: MFCD11855994
CAS Number: 179024-68-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR