Methyl-4,8-dichloroquinoline-2-carboxylate
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HC-2319-1G
HC-2319-5G
COBBHC-2319-1GEA
0
GBP
COBBHC-2319-1G
COBBHC-2319-5G
Methyl-4,8-dichloroquinoline-2-carboxylate
Methyl 4,8-dichloroquinoline-2-carboxylate
Formula:
C₁₁H₇Cl₂NO₂ |
MDL Number:
MFCD11855883 CAS Number: 1020101-15-8 |
Learn more
![](/m-uk.vwr.com/en_GB/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...