Methyl-4,6-dichloroquinoline-2-carboxylate

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-2320-1G HC-2320-5G
COBBHC-2320-1GEA 0 GBP
COBBHC-2320-1G COBBHC-2320-5G
Methyl-4,6-dichloroquinoline-2-carboxylate
Methyl 4,6-dichloroquinoline-2-carboxylate
Formula: C₁₁H₇Cl₂NO₂
MDL Number: MFCD08458388
CAS Number: 848501-96-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR