8-Bromo-5-nitroquinoline

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-2060-5G HC-2060-1G
COBBHC-2060-5GEA 0 GBP
COBBHC-2060-5G COBBHC-2060-1G
8-Bromo-5-nitroquinoline
8-Bromo-5-nitroquinoline
Formula: C₉H₅BrN₂O₂
MW: 253.0500 g/mol
MDL Number: MFCD00509069
CAS Number: 139366-35-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR