8-Bromo-5-(trifluoromethoxy)quinoline

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-2439-1G HC-2439-250M
COBBHC-2439-1GEA 0 GBP
COBBHC-2439-1G COBBHC-2439-250M
8-Bromo-5-(trifluoromethoxy)quinoline
8-Bromo-5-(trifluoromethoxy)quinoline
Formula: C₁₀H₅BrF₃NO
MDL Number: MFCD11855905
CAS Number: 1133115-91-9

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR