Methyl-8-bromo-4,6-dichloroquinoline-2-carboxylate
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HC-2541-5G
HC-2541-1G
COBBHC-2541-5GEA
0
GBP
COBBHC-2541-5G
COBBHC-2541-1G
Methyl-8-bromo-4,6-dichloroquinoline-2-carboxylate
Methyl 8-bromo-4,6-dichloroquinoline-2-carboxylate
Formula:
C₁₁H₆BrCl₂NO₂ |
MDL Number:
MFCD12026025 CAS Number: 1150164-76-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...