6-Bromo-4-chloroquinoline

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS5057G1 AS5057G25 AS5057G5
ACTIAS5057G1EA 0 GBP
ACTIAS5057G1 ACTIAS5057G25 ACTIAS5057G5
6-Bromo-4-chloroquinoline
6-Bromo-4-chloroquinoline
Formula: C₉H₅BrClN
MW: 242.50 g/mol
MDL Number: MFCD00511001
CAS Number: 65340-70-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR