6-Bromo-2-chloroquinoline

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS2243G1 AS2243G25 AS2243G5
ACTIAS2243G1EA 0 GBP
ACTIAS2243G1 ACTIAS2243G25 ACTIAS2243G5
6-Bromo-2-chloroquinoline
6-Bromo-2-chloroquinoline
Formula: C₉H₅BrClN
MDL Number: MFCD04115272
CAS Number: 1810-71-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR